missing translation for 'onlineSavingsMsg'
Learn More

9-Nitroanthracene 90.0+%, TCI America™

Catalog No. N09875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09875G 5 g
N09871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09875G Supplier TCI America Supplier No. N09875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 602-60-8
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00001248
InChI Key LSIKFJXEYJIZNB-UHFFFAOYSA-N
Synonym anthracene, 9-nitro, 5-nitroanthracene, unii-jz1tb3n57p, 9-nitro-anthracene, ccris 4679, jz1tb3n57p, 9-nitro anthracene, pubchem15119, acmc-20ab5c, 9-nitroanthracene
PubChem CID 11767
ChEBI CHEBI:82501
IUPAC Name 9-nitroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2[N+](=O)[O-]

Specifications

Melting Point 144°C
Boiling Point 275°C
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Nitroanthracene
RTECSNumber CB0715000
TSCA No