missing translation for 'onlineSavingsMsg'
Learn More

9-Methylanthracene 98.0+%, TCI America™

Catalog No. p-7132021
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M01531G 1 g
M01535G 5 g
2 options

Catalog No. M01531G

Supplier: TCI America M01531G

Only null left

Chemical Identifiers

CAS 779-02-2
Molecular Formula C15H12
Molecular Weight (g/mol) 192.26
MDL Number MFCD00001261
InChI Key CPGPAVAKSZHMBP-UHFFFAOYSA-N
Synonym anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
PubChem CID 13068
IUPAC Name 9-methylanthracene
SMILES CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 79°C
Boiling Point 370°C
Quantity 1 g
UN Number 3077
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Methylanthracene
EINECSNumber (4)-0640
RTECSNumber CB0700000
TSCA Yes