accessibility menu, dialog, popup

UserName

CAS RN 779-02-2

IUPAC Name: 9-methylanthracene
Synonyms: 9-methylanthracene
Molecular Weight (g/mol): 192.26
Molecular Formula: C15H12
InChi Key: CPGPAVAKSZHMBP-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=CC2=CC=CC=C12
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)