Learn More
9-Methylanthracene, 99%
CAS: 779-02-2 | C15H12 | 192.26 g/mol
Supplier: Thermo Scientific Chemicals A1569906
Description
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.
Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.
Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
Specifications
| 9-Methylanthracene | |
| 77°C to 79°C | |
| 196°C to 197°C (16 mmHg) | |
| MFCD00001261 | |
| UN3077 | |
| anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard | |
| CPGPAVAKSZHMBP-UHFFFAOYSA-N | |
| 9-methylanthracene | |
| 13068 | |
| 99% |
| 779-02-2 | |
| 1.066 | |
| C15H12 | |
| 5 g | |
| 1907463 | |
| Soluble in water (0,0003mg/L at 25°C) and toluene. Slightly soluble in benzene. | |
| CC1=C2C=CC=CC2=CC2=CC=CC=C12 | |
| 192.26 | |
| 192.26 |
Chemical Identifiers
| 779-02-2 | |
| 192.26 | |
| CPGPAVAKSZHMBP-UHFFFAOYSA-N | |
| 13068 | |
| CC1=C2C=CC=CC2=CC2=CC=CC=C12 |
| C15H12 | |
| MFCD00001261 | |
| anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard | |
| 9-methylanthracene |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 212-299-7
RTECSNumber : CB0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only