missing translation for 'onlineSavingsMsg'
Learn More

9-Methyl-9H-fluoren-9-ol 98.0+%, TCI America™

Catalog No. p-7133532
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30071G 1 g
M30075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30071G Supplier TCI America Supplier No. M30071G

Chemical Identifiers

CAS 6311-22-4
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00183690
InChI Key ZMXJQEIJNHMYDY-UHFFFAOYSA-N
Synonym 9-Hydroxy-9-methyl-9H-fluorene
PubChem CID 95933
IUPAC Name 9-methyl-9H-fluoren-9-ol
SMILES CC1(O)C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Methyl-9H-fluoren-9-ol