accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 6311-22-4

IUPAC Name: 9-methyl-9H-fluoren-9-ol
Synonyms: 9-methylfluoren-9-ol
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: ZMXJQEIJNHMYDY-UHFFFAOYSA-N
SMILES: CC1(O)C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results