missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl Carbamate 98.0+%, TCI America™

Catalog No. F06891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06891G 1 g
F06895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06891G Supplier TCI America Supplier No. F06891G

Chemical Identifiers

CAS 84418-43-9
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
MDL Number MFCD00270532
InChI Key ZZOKVYOCRSMTSS-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl carbamate, fmoc-nh2, fmoc-amide, 9-fluorenylmethylcarbamate, n-9-fluorenylmethoxycarbonyl amide, fmoc amine, fmoc-amine, 9-fluorenylcarbamate, 9-fluorenemethane resin, pubchem12066
PubChem CID 736301
IUPAC Name 9H-fluoren-9-ylmethyl carbamate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 1 g
Formula Weight 239.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenylmethyl Carbamate