accessibility menu, dialog, popup

UserName

OOH2N

CAS RN 84418-43-9

IUPAC Name: (9H-fluoren-9-yl)methyl carbamate
Synonyms: 9H-fluoren-9-ylmethyl carbamate
Molecular Weight (g/mol): 239.27
Molecular Formula: C15H13NO2
InChi Key: ZZOKVYOCRSMTSS-UHFFFAOYSA-N
SMILES: NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)