missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl carbamate, 99%

Catalog No. AC434170010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434170010 1 g Glass bottle
AC434170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434170010 Supplier Thermo Scientific Chemicals Supplier No. 434170010

CAS: 84418-43-9 | C15H13NO2 | 239.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84418-43-9
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.28
MDL Number MFCD00237376
InChI Key ZZOKVYOCRSMTSS-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl carbamate, fmoc-nh2, fmoc-amide, 9-fluorenylmethylcarbamate, n-9-fluorenylmethoxycarbonyl amide, fmoc amine, fmoc-amine, 9-fluorenylcarbamate, 9-fluorenemethane resin, pubchem12066
PubChem CID 736301
IUPAC Name 9H-fluoren-9-ylmethyl carbamate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 204°C to 207°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 239.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 9-Fluorenylmethyl carbamate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only