missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone-4-carboxylic acid, 98+%

Catalog No. AAB2502103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2502103 1 g
AAB2502106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2502103 Supplier Thermo Scientific Chemicals Supplier No. B2502103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6223-83-2 | C14H8O3 | 224.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6223-83-2
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.215
MDL Number MFCD00001145
InChI Key AFQYQSWTVCNJQT-UHFFFAOYSA-N
Synonym 9-fluorenone-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid, 9h-fluorene-4-carboxylic acid, 9-oxo, 9-fluorenone-4-carboxlic acid, 9-oxofluorene-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid #
PubChem CID 80361
IUPAC Name 9-oxofluorene-4-carboxylic acid
SMILES C1=CC=C2C(=C1)C3=C(C2=O)C=CC=C3C(=O)O

Specifications

Melting Point 224°C to 228°C
Quantity 1 g
Beilstein 2694801
Formula Weight 224.22
Percent Purity ≥98%
Chemical Name or Material 9-Fluorenone-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-311-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only