accessibility menu, dialog, popup

UserName

CAS RN 6223-83-2

IUPAC Name: 9-oxo-9H-fluorene-4-carboxylic acid
Synonyms: 9-oxofluorene-4-carboxylic acid
Molecular Weight (g/mol): 224.22
Molecular Formula: C14H8O3
InChi Key: AFQYQSWTVCNJQT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)