missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenol 99.0+%, TCI America™

Catalog No. p-7129555
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F022925G 25 g
F0229250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F022925G Supplier TCI America Supplier No. F022925G
Only null left

Chemical Identifiers

CAS 1689-64-1
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00001135
InChI Key AFMVESZOYKHDBJ-UHFFFAOYSA-N
Synonym 9-hydroxyfluorene, 9-fluorenol, fluoren-9-ol, fluorenol, 9-hydroxy fluorene, diphenylene carbinol, unii-bv0q72r613, hydroxyfluoren, 9-hydroxy-fluoren, 9h-fluorene-9-ol
PubChem CID 74318
ChEBI CHEBI:16904
IUPAC Name 9H-fluoren-9-ol
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 182.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenol