missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenecarboxylic Acid 97.0+%, TCI America™

Supplier:  TCI America F002225G

 View more versions of this product

Catalog No. F002225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

9-Fluorenecarboxylic Acid
230°C
C14H10O2
25 g
DNVJGJUGFFYUPT-UHFFFAOYSA-N
9H-fluorene-9-carboxylic acid
74809
≥97.0% (T)
1989-33-9
White-Yellow
MFCD00001136
fluorene-9-carboxylic acid, 9-fluorenecarboxylic acid, 9-carboxyfluorene, diphenyleneacetic acid, 9-fluorencarboxylic acid, rarechem aq bd 0ba1, akos auf02021, 9-carboxy-9h-fluorene, enamine_003081, 9-fluorenylcarboxylic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)C(=O)O
210.232
210.23
Crystalline Powder

Chemical Identifiers

1989-33-9
210.232
DNVJGJUGFFYUPT-UHFFFAOYSA-N
74809
C1=CC=C2C(=C1)C(C3=CC=CC=C32)C(=O)O
C14H10O2
MFCD00001136
fluorene-9-carboxylic acid, 9-fluorenecarboxylic acid, 9-carboxyfluorene, diphenyleneacetic acid, 9-fluorencarboxylic acid, rarechem aq bd 0ba1, akos auf02021, 9-carboxy-9h-fluorene, enamine_003081, 9-fluorenylcarboxylic acid
9H-fluorene-9-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions