missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenecarboxylic Acid 97.0+%, TCI America™

Catalog No. p-7129445
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F00225G 5 g
F002225G 25 g
2 options

Catalog No. F00225G

Supplier: TCI America F00225G

Only null left

Chemical Identifiers

CAS 1989-33-9
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
MDL Number MFCD00001136
InChI Key DNVJGJUGFFYUPT-UHFFFAOYSA-N
Synonym fluorene-9-carboxylic acid, 9-fluorenecarboxylic acid, 9-carboxyfluorene, diphenyleneacetic acid, 9-fluorencarboxylic acid, rarechem aq bd 0ba1, akos auf02021, 9-carboxy-9h-fluorene, enamine_003081, 9-fluorenylcarboxylic acid
PubChem CID 74809
IUPAC Name 9H-fluorene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)C(=O)O

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenecarboxylic Acid