missing translation for 'onlineSavingsMsg'
Learn More

9-Fluoreneacetic Acid 98.0+%, TCI America™

Catalog No. p-7129617
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
F03301G 1 g
F03305G 5 g
2 options

Catalog No. F03301G

Supplier: TCI America F03301G

Only null left

Chemical Identifiers

CAS 6284-80-6
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00013262
InChI Key WFSMJMTYIMFHPV-UHFFFAOYSA-N
Synonym 9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid
PubChem CID 94852
IUPAC Name 2-(9H-fluoren-9-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 224.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluoreneacetic Acid