missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene, 96%

Catalog No. AAL1422506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1422506 5 g
AAL1422503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1422506 Supplier Thermo Scientific Chemicals Supplier No. L1422506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55135-66-5 | C19H13Br | 321.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55135-66-5
Molecular Formula C19H13Br
Molecular Weight (g/mol) 321.217
MDL Number MFCD00075522
InChI Key HYQXNCDBSALQLB-UHFFFAOYSA-N
Synonym 9-bromo-9-phenyl-9h-fluorene, 9-brom-9-phenyl-9h-fluoren, zlchem 855, acmc-209ll7, 9-bromo-9-phenyl-fluorene, 9-bromanyl-9-phenyl-fluorene, hyqxncdbsalqlb-uhfffaoysa, 9-bromo-9-phenylfluorene, 9h-fluorene, 9-bromo-9-phenyl
PubChem CID 231624
IUPAC Name 9-bromo-9-phenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)Br

Specifications

Melting Point 94°C to 99°C
Quantity 5 g
UN Number UN3261
Beilstein 2561445
Formula Weight 321.23
Percent Purity 96%
Chemical Name or Material 9-Bromo-9-phenylfluorene
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only