missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Bromo-10-(9-phenanthryl)anthracene 98.0+%, TCI America™
Supplier: TCI America B45721G
Specifications
| 9-Bromo-10-(9-phenanthryl)anthracene | |
| 232°C | |
| MFCD27939628 | |
| OBIXPXSRUDGFLE-UHFFFAOYSA-N | |
| 9-bromo-10-(phenanthren-9-yl)anthracene | |
| 86031959 | |
| ≥98.0% (HPLC) |
| 845457-53-6 | |
| C28H17Br | |
| 1 g | |
| BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12 | |
| 433.35 | |
| 433.35 | |
| Crystalline Powder |
Chemical Identifiers
| 845457-53-6 | |
| 433.35 | |
| OBIXPXSRUDGFLE-UHFFFAOYSA-N | |
| 9-bromo-10-(phenanthren-9-yl)anthracene |
| C28H17Br | |
| MFCD27939628 | |
| 86031959 | |
| BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
TSCA : No