missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-(9-phenanthryl)anthracene 98.0+%, TCI America™

Catalog No. p-7123627
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4572200MG 200 mg
B45721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4572200MG Supplier TCI America Supplier No. B4572200MG
Only null left

Chemical Identifiers

CAS 845457-53-6
Molecular Formula C28H17Br
Molecular Weight (g/mol) 433.35
MDL Number MFCD27939628
InChI Key OBIXPXSRUDGFLE-UHFFFAOYSA-N
PubChem CID 86031959
IUPAC Name 9-bromo-10-(phenanthren-9-yl)anthracene
SMILES BrC1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 232°C
Quantity 200 mg
Formula Weight 433.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Bromo-10-(9-phenanthryl)anthracene