missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 468635000

 View more versions of this product

Catalog No. AC468635000


Only null left

Description

Description

Specifications

Specifications

9-beta-D-Arabinofuranosylguanine
Available
Crystalline Powder
MFCD00065486
225°C
NYHBQMYGNKIUIF-FJFJXFQQSA-N
2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
500 mg
Glass Bottle
38819-10-2
C10H13N5O5
White
Araguanosine; Ara-G
NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
283.24
283.24

Chemical Identifiers

38819-10-2
283.24
NYHBQMYGNKIUIF-FJFJXFQQSA-N
2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
C10H13N5O5
MFCD00065486
Araguanosine; Ara-G
NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS08: Health hazard

GHS07: Exclamation mark

ShelfLife : 3 years

RTECSNumber : MF8301000

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Promotions

Promotions