missing translation for 'onlineSavingsMsg'
Learn More

9-beta-D-Arabinofuranosylguanine, Thermo Scientific™

Catalog No. p-7232411
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468631000 100 mg
AC468635000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468631000 Supplier Thermo Scientific Chemicals Supplier No. 468631000
Only null left

Chemical Identifiers

CAS 38819-10-2
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00065486
InChI Key NYHBQMYGNKIUIF-FJFJXFQQSA-N
Synonym Araguanosine; Ara-G
IUPAC Name 2-amino-9-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White
Melting Point 225°C
Quantity 100 mg
Formula Weight 283.24
Chemical Name or Material 9-beta-D-Arabinofuranosylguanine
Hazard Category GHS07: Exclamation mark
Hazard Statement GHS08: Health hazard
ShelfLife 3 years
RTECSNumber MF8301000
Recommended Storage Refrigerator +4°C