missing translation for 'onlineSavingsMsg'
Learn More

9-Anthracenemethanol, 97%

CAS: 1468-95-7 | C15H12O | 208.26 g/mol

Supplier:  Thermo Scientific Chemicals A1518818

 View more versions of this product

Catalog No. AAA1518818


Only null left
Explore available promotions

Description

Description

An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

9-Anthracenemethanol
160°C to 165°C
MFCD00001264
1873402
Soluble in hot methanol very faint turbidity.
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
208.26
208.26
1468-95-7
C15H12O
50 g
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
JCJNNHDZTLRSGN-UHFFFAOYSA-N
anthracen-9-ylmethanol
73848
97%

Chemical Identifiers

1468-95-7
208.26
JCJNNHDZTLRSGN-UHFFFAOYSA-N
73848
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
C15H12O
MFCD00001264
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
anthracen-9-ylmethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-998-5

RTECSNumber : CB0577500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only