missing translation for 'onlineSavingsMsg'
Learn More

9-Aminofluorene hydrochloride, 98+%

Catalog No. AAL0515603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0515603 1 g
AAL0515606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0515603 Supplier Thermo Scientific Chemicals Supplier No. L0515603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5978-75-6 | C13H12ClN | 217.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5978-75-6
Molecular Formula C13H12ClN
Molecular Weight (g/mol) 217.70
MDL Number MFCD00012536
InChI Key SYKJOJSYQSVNOM-UHFFFAOYSA-N
Synonym 9h-fluoren-9-amine hydrochloride, 9-aminofluorene hydrochloride, 9h-fluoren-9-amine, hydrochloride, 9-fluorenamine hydrochloride, 9-aminofluorene hcl, fluorene-9-ylamine, chloride, 9h-fluoren-9-amine hcl, acmc-1b10k, 9-aminoflu-orene hydrochloride, 9-aminofluorene monohydrochloride
PubChem CID 2724685
SMILES [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point ∼225°C
Quantity 1 g
Beilstein 3915368
Sensitivity Hygroscopic
IUPAC Name 9H-fluoren-9-amine;hydrochloride
Formula Weight 217.7
Percent Purity ≥98%
Chemical Name or Material 9-Aminofluorene hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 227-778-6
RTECSNumber LL5130000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only