missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Aminoanthracene 96.0+%, TCI America™
$169.63 - $2486.65
Chemical Identifiers
| CAS | 779-03-3 |
|---|---|
| Molecular Formula | C14H11N |
| Molecular Weight (g/mol) | 193.249 |
| MDL Number | MFCD00452690 |
| InChI Key | LHNICELDCMPPDE-UHFFFAOYSA-N |
| Synonym | 9-Anthramine |
| PubChem CID | 13069 |
| ChEBI | CHEBI:50472 |
| IUPAC Name | anthracen-9-amine |
| SMILES | C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1748100MG
|
TCI America
A1748100MG |
100 mg |
Each for $169.63
|
|
|||||
|
A17481G
|
TCI America
A17481G |
1 g |
Each for $831.37
|
|
|||||
|
A17485G
|
TCI America
A17485G |
5 g |
Each for $2,486.65
|
|
|||||
Chemical Identifiers
| 779-03-3 | |
| 193.249 | |
| LHNICELDCMPPDE-UHFFFAOYSA-N | |
| 13069 | |
| anthracen-9-amine |
| C14H11N | |
| MFCD00452690 | |
| 9-Anthramine | |
| CHEBI:50472 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N |
Specifications
| 779-03-3 | |
| Green-Yellow | |
| MFCD00452690 | |
| 9-Anthramine | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N | |
| 193.249 | |
| CHEBI:50472 | |
| ≥96.0% (GC,N) | |
| 9-Aminoanthracene |
| 170°C | |
| C14H11N | |
| 100 mg | |
| LHNICELDCMPPDE-UHFFFAOYSA-N | |
| anthracen-9-amine | |
| 13069 | |
| 193.25 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CB2279000
TSCA : No