missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoanthracene 96.0+%, TCI America™

Supplier:  TCI America A17481G

 View more versions of this product

Catalog No. A17481G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

9-Aminoanthracene
170°C
C14H11N
1 g
LHNICELDCMPPDE-UHFFFAOYSA-N
anthracen-9-amine
13069
193.25
Crystalline Powder
779-03-3
Green-Yellow
MFCD00452690
9-Anthramine
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N
193.249
CHEBI:50472
≥96.0% (GC,N)

Chemical Identifiers

779-03-3
193.249
LHNICELDCMPPDE-UHFFFAOYSA-N
13069
anthracen-9-amine
C14H11N
MFCD00452690
9-Anthramine
CHEBI:50472
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CB2279000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions