missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoacridine hemihydrate, 98%

Catalog No. AC134410010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC134410010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC134410010 Supplier Thermo Scientific Chemicals Supplier No. 134410010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65944-23-2 | C13H10N2·1/2H2O | 203.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65944-23-2 , 90-45-9
Molecular Formula C13H10N2·1/2H2O
Molecular Weight (g/mol) 203.16
MDL Number MFCD00150520
InChI Key SPWFQHCZHQCDAG-UHFFFAOYSA-N
Synonym acridin-9-amine hydrate, 9-aminoacridine hemihydrate
PubChem CID 2723763
IUPAC Name acridin-9-amine;hydrate
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C to 235.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 21, II, 280
Merck Index 14, 407
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,freely soluble in alcohol,slightly soluble in chloroform,toluene and,pyridine
Formula Weight 203.16
Percent Purity 98%
Chemical Name or Material 9-Aminoacridine hemihydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get

RUO – Research Use Only