accessibility menu, dialog, popup

UserName

CAS RN 90-45-9

IUPAC Name: acridin-9-amine
Synonyms: 9-aminoacridine
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: XJGFWWJLMVZSIG-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results