CAS RN 65944-23-2
IUPAC Name:
bis(acridin-9-amine) hydrate
Synonyms:
bis(9-aminoacridine) hydrate
Molecular Weight (g/mol):
406.49
Molecular Formula:
C26H22N4O
InChi Key:
SPWFQHCZHQCDAG-UHFFFAOYSA-N
SMILES:
O.NC1=C2C=CC=CC2=NC2=CC=CC=C12.NC1=C2C=CC=CC2=NC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
9-Aminoacridine hemihydrate, 98%
9-Aminoacridine hemihydrate, 98%
CAS: 65944-23-2 Molecular Formula: C13H10N2·1/2H2O Molecular Weight (g/mol): 203.16 MDL Number: MFCD00150520 InChI Key: SPWFQHCZHQCDAG-UHFFFAOYSA-N Synonym: acridin-9-amine hydrate,9-aminoacridine hemihydrate PubChem CID: 2723763 IUPAC Name: acridin-9-amine;hydrate SMILES: C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.O