Learn More
9-Acetylanthracene, 96%
CAS: 784-04-3 | C16H12O | 220.271 g/mol
Supplier: Thermo Scientific Chemicals L0127806
Description
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
Specifications
| 9-Acetylanthracene | |
| 72°C to 76°C | |
| MFCD00001259 | |
| 1370056 | |
| Insoluble in water. | |
| CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31 | |
| 220.271 | |
| 220.27 |
| 784-04-3 | |
| C16H12O | |
| 5 g | |
| 9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene | |
| NXXNVJDXUHMAHU-UHFFFAOYSA-N | |
| 1-anthracen-9-ylethanone | |
| 69911 | |
| 96% |
Chemical Identifiers
| 784-04-3 | |
| 220.271 | |
| NXXNVJDXUHMAHU-UHFFFAOYSA-N | |
| 69911 | |
| CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31 |
| C16H12O | |
| MFCD00001259 | |
| 9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene | |
| 1-anthracen-9-ylethanone |
Safety and Handling
EINECSNumber : 212-315-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only