missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Acetylanthracene, 96%
Description
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Insoluble in water.
Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 72°C to 76°C |
| Quantity | 5 g |
| Beilstein | 1370056 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 220.27 |
| Percent Purity | 96% |
| Chemical Name or Material | 9-Acetylanthracene |
Safety and Handling
| EINECSNumber | 212-315-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only