missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobifluorene, 98%

CAS: 159-66-0 | C25H16 | 316.403 g/mol

Supplier:  Thermo Scientific Chemicals H6413303

 View more versions of this product

Catalog No. AAH6413303


Only null left
Explore available promotions

Description

Description

A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

9,9'-Spirobifluorene
195°C to 200°C
C25H16
1 g
9,9'-spirobifluorene, 9,9'-spirobi fluorene, 9,9'-spirobi 9h-fluorene, 9,9-spirobi 9h-fluorene, 9,9-spirobifluorene, pubchem3102, 9.9'-spirobifluorene, acmc-209dk2, 9,9 inverted exclamation mark-spirobi-fluorene
SNFCXVRWFNAHQX-UHFFFAOYSA-N
9,9'-spirobi[fluorene]
135975
98%
159-66-0
White to Yellow
MFCD08704216
1914463
Soluble in common organic solvents. Slightly soluble in water.
C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46
316.403
316.4
Solid

Chemical Identifiers

159-66-0
316.403
SNFCXVRWFNAHQX-UHFFFAOYSA-N
135975
C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46
C25H16
MFCD08704216
9,9'-spirobifluorene, 9,9'-spirobi fluorene, 9,9'-spirobi 9h-fluorene, 9,9-spirobi 9h-fluorene, 9,9-spirobifluorene, pubchem3102, 9.9'-spirobifluorene, acmc-209dk2, 9,9 inverted exclamation mark-spirobi-fluorene
9,9'-spirobi[fluorene]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only