accessibility menu, dialog, popup

UserName

CAS RN 159-66-0

IUPAC Name: 9,9'-spirobi[fluorene]
Synonyms: 9,9'-spirobi[fluorene]
Molecular Weight (g/mol): 316.4
Molecular Formula: C25H16
InChi Key: SNFCXVRWFNAHQX-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)