missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobifluorene, 98%

Catalog No. AAH64133MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64133MD 250 mg
AAH6413303 1 g
AAH6413306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64133MD Supplier Thermo Scientific Chemicals Supplier No. H64133MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 159-66-0 | C25H16 | 316.403 g/mol

A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 159-66-0
Molecular Formula C25H16
Molecular Weight (g/mol) 316.403
MDL Number MFCD08704216
InChI Key SNFCXVRWFNAHQX-UHFFFAOYSA-N
Synonym 9,9'-spirobifluorene, 9,9'-spirobi fluorene, 9,9'-spirobi 9h-fluorene, 9,9-spirobi 9h-fluorene, 9,9-spirobifluorene, pubchem3102, 9.9'-spirobifluorene, acmc-209dk2, 9,9 inverted exclamation mark-spirobi-fluorene
PubChem CID 135975
IUPAC Name 9,9'-spirobi[fluorene]
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 195°C to 200°C
Color White to Yellow
Quantity 250 mg
Beilstein 1914463
Solubility Information Soluble in common organic solvents. Slightly soluble in water.
Formula Weight 316.4
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9,9'-Spirobifluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only