missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobi[9H-fluorene] 98.0+%, TCI America™

Catalog No. p-7135937
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08001G 1 g
S08005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08001G Supplier TCI America Supplier No. S08001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 159-66-0
Molecular Formula C25H16
Molecular Weight (g/mol) 316.403
MDL Number MFCD08704216
InChI Key SNFCXVRWFNAHQX-UHFFFAOYSA-N
Synonym 9,9'-spirobifluorene, 9,9'-spirobi fluorene, 9,9'-spirobi 9h-fluorene, 9,9-spirobi 9h-fluorene, 9,9-spirobifluorene, pubchem3102, 9.9'-spirobifluorene, acmc-209dk2, 9,9 inverted exclamation mark-spirobi-fluorene
PubChem CID 135975
IUPAC Name 9,9'-spirobi[fluorene]
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 1 g
Formula Weight 316.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-Spirobi[9H-fluorene]