missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Dimethyl-2-nitrofluorene 98.0+%, TCI America™
$91.33 - $315.03
Chemical Identifiers
| CAS | 605644-46-0 |
|---|---|
| Molecular Formula | C15H13NO2 |
| Molecular Weight (g/mol) | 239.274 |
| MDL Number | MFCD17170053 |
| InChI Key | SPNVINZCDHRVAI-UHFFFAOYSA-N |
| PubChem CID | 46912048 |
| IUPAC Name | 9,9-dimethyl-2-nitrofluorene |
| SMILES | CC1(C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-])C |
Chemical Identifiers
| 605644-46-0 | |
| 239.274 | |
| SPNVINZCDHRVAI-UHFFFAOYSA-N | |
| 9,9-dimethyl-2-nitrofluorene |
| C15H13NO2 | |
| MFCD17170053 | |
| 46912048 | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-])C |
Specifications
| 605644-46-0 | |
| White-Yellow | |
| MFCD17170053 | |
| SPNVINZCDHRVAI-UHFFFAOYSA-N | |
| 9,9-dimethyl-2-nitrofluorene | |
| 46912048 | |
| ≥98.0% (GC) | |
| 9,9-Dimethyl-2-nitrofluorene |
| 103°C | |
| C15H13NO2 | |
| 5 g | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-])C | |
| 239.274 | |
| 239.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No