missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Dimethyl-2-nitrofluorene 98.0+%, TCI America™
Supplier: TCI America D460925G
Specifications
| 9,9-Dimethyl-2-nitrofluorene | |
| 103°C | |
| C15H13NO2 | |
| 25 g | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-])C | |
| 239.274 | |
| 239.27 | |
| Crystalline Powder |
| 605644-46-0 | |
| White-Yellow | |
| MFCD17170053 | |
| SPNVINZCDHRVAI-UHFFFAOYSA-N | |
| 9,9-dimethyl-2-nitrofluorene | |
| 46912048 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 605644-46-0 | |
| 239.274 | |
| SPNVINZCDHRVAI-UHFFFAOYSA-N | |
| 9,9-dimethyl-2-nitrofluorene |
| C15H13NO2 | |
| MFCD17170053 | |
| 46912048 | |
| CC1(C2=CC=CC=C2C3=C1C=C(C=C3)[N+](=O)[O-])C |
Safety and Handling
TSCA : No