missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di(p-tolyl)fluorene 98.0+%, TCI America™

Catalog No. D46101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46101G 1 g
D46105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46101G Supplier TCI America Supplier No. D46101G
Only null left

Chemical Identifiers

CAS 54941-50-3
Molecular Formula C27H22
Molecular Weight (g/mol) 346.47
MDL Number MFCD23115707
InChI Key ARZDANJGUJGWIO-UHFFFAOYSA-N
PubChem CID 23369087
IUPAC Name 9,9-bis(4-methylphenyl)-9H-fluorene
SMILES CC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(C)C=C1

Specifications

Melting Point 169°C
Color White
Quantity 1 g
Formula Weight 346.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Di(p-tolyl)fluorene