accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 54941-50-3

IUPAC Name: 9,9-bis(4-methylphenyl)-9H-fluorene
Synonyms: 9,9-bis(4-methylphenyl)fluorene
Molecular Weight (g/mol): 346.47
Molecular Formula: C27H22
InChi Key: ARZDANJGUJGWIO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results