missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7052786
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5581703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5581703 Supplier Thermo Scientific Chemicals Supplier No. H5581703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 203927-98-4 | Molecular Formula: C25H36B2O4 | Molecular Weight: 422.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 203927-98-4
Molecular Formula C25H36B2O4
Molecular Weight (g/mol) 422.18
MDL Number MFCD03427279
InChI Key QFWOJNOZWMHNTK-UHFFFAOYSA-N
Synonym 9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene
PubChem CID 4524285
SMILES CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O

Specifications

Quantity 1 g
IUPAC Name [7-(dihydroxyboranyl)-9,9-dihexyl-9H-fluoren-2-yl]boronic acid
Formula Weight 422.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 9,9-Di-n-hexylfluorene-2,7-diboronic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only