missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%

Catalog No. AAH5632014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5632014 25 g
AAH5632022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5632014 Supplier Thermo Scientific Chemicals Supplier No. H5632014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 286438-45-7 | C37H56Br2 | 660.663 g/mol

It is an intermediate for polymeric light-emitting diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate for polymeric light-emitting diodes.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 286438-45-7
Molecular Formula C37H56Br2
Molecular Weight (g/mol) 660.663
MDL Number MFCD03427215
InChI Key KFOUJVGPGBSIFB-UHFFFAOYSA-N
Synonym 2,7-dibromo-9,9-didodecyl-9h-fluorene, 9,9-didodecyl-2,7-dibromofluorene, 9 9-didodecyl-2 7-dibromofluorene, 9h-fluorene, 2,7-dibromo-9,9-didodecyl, 2,7-dibromo-9,9-didodecyl-fluorene, acmc-209h3v, ksc561q2d, 2,7-dibromo-didodecyl-9h-fluorene, 9,9-di-n-dodecyl-2,7-dibromofluorene, 9h-fluorene,2,7-dibromo-9,9-didodecyl
PubChem CID 5140914
IUPAC Name 2,7-dibromo-9,9-didodecylfluorene
SMILES CCCCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCCCC

Specifications

Melting Point 50°C to 55°C
Boiling Point 180°C (0.15 mmHg)
Flash Point 110°C (230°F)
Quantity 25 g
Formula Weight 660.65
Percent Purity 98%
Chemical Name or Material 9,9-Di-n-dodecyl-2,7-dibromofluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only