missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Bis(4-bromophenyl)fluorene 98.0+%, TCI America™
$283.00 - $958.74
Chemical Identifiers
| CAS | 128406-10-0 |
|---|---|
| Molecular Formula | C25H16Br2 |
| Molecular Weight (g/mol) | 476.211 |
| InChI Key | VQTLUEKUYMRVDS-UHFFFAOYSA-N |
| PubChem CID | 10695915 |
| IUPAC Name | 9,9-bis(4-bromophenyl)fluorene |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)Br)C5=CC=C(C=C5)Br |
Chemical Identifiers
| 128406-10-0 | |
| 476.211 | |
| 10695915 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)Br)C5=CC=C(C=C5)Br |
| C25H16Br2 | |
| VQTLUEKUYMRVDS-UHFFFAOYSA-N | |
| 9,9-bis(4-bromophenyl)fluorene |
Specifications
| 128406-10-0 | |
| White | |
| 1 g | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)Br)C5=CC=C(C=C5)Br | |
| 476.211 | |
| 476.21 | |
| Crystalline Powder |
| 158°C | |
| C25H16Br2 | |
| VQTLUEKUYMRVDS-UHFFFAOYSA-N | |
| 9,9-bis(4-bromophenyl)fluorene | |
| 10695915 | |
| ≥98.0% (GC) | |
| 9,9-Bis(4-bromophenyl)fluorene |
Safety and Handling
TSCA : No