accessibility menu, dialog, popup

UserName

BrBr

CAS RN 128406-10-0

IUPAC Name: 9,9-bis(4-bromophenyl)-9H-fluorene
Synonyms: 9,9-bis(4-bromophenyl)fluorene
Molecular Weight (g/mol): 476.21
Molecular Formula: C25H16Br2
InChi Key: VQTLUEKUYMRVDS-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results