missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-amino-3-fluorophenyl)fluorene 98.0+%, TCI America™

Catalog No. B26915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26915G Supplier TCI America Supplier No. B26915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127926-65-2
Molecular Formula C25H18F2N2
Molecular Weight (g/mol) 384.43
InChI Key RXNKCIBVUNMMAD-UHFFFAOYSA-N
Synonym 2,2′C-Difluoro-4,4′C-(9-fluorenylidene)dianiline
PubChem CID 44629774
IUPAC Name 4-[9-(4-amino-3-fluorophenyl)fluoren-9-yl]-2-fluoroaniline
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC(=C(C=C4)N)F)C5=CC(=C(C=C5)N)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 384.43
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-amino-3-fluorophenyl)fluorene