accessibility menu, dialog, popup

UserName

H2NFNH2F

CAS RN 127926-65-2

IUPAC Name: 4-[9-(4-amino-3-fluorophenyl)-9H-fluoren-9-yl]-2-fluoroaniline
Synonyms: 4-[9-(4-amino-3-fluorophenyl)fluoren-9-yl]-2-fluoroaniline
Molecular Weight (g/mol): 384.43
Molecular Formula: C25H18F2N2
InChi Key: RXNKCIBVUNMMAD-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(F)=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results