missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-amino-3-chlorophenyl)fluorene 98.0+%, TCI America™

Catalog No. B26905G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26905G Supplier TCI America Supplier No. B26905G
Only null left

Chemical Identifiers

CAS 107934-68-9
Molecular Formula C25H18Cl2N2
Molecular Weight (g/mol) 417.333
InChI Key CIZUMWWHWPJAAK-UHFFFAOYSA-N
Synonym 2,2′C-Dichloro-4,4′C-(9-fluorenylidene)dianiline
PubChem CID 21872159
IUPAC Name 4-[9-(4-amino-3-chlorophenyl)fluoren-9-yl]-2-chloroaniline
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC(=C(C=C4)N)Cl)C5=CC(=C(C=C5)N)Cl

Specifications

Melting Point 200°C
Quantity 5 g
UN Number 3077
Formula Weight 417.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-amino-3-chlorophenyl)fluorene