accessibility menu, dialog, popup

UserName

H2NClNH2Cl

CAS RN 107934-68-9

IUPAC Name: 4-[9-(4-amino-3-chlorophenyl)-9H-fluoren-9-yl]-2-chloroaniline
Synonyms: 4-[9-(4-amino-3-chlorophenyl)fluoren-9-yl]-2-chloroaniline
Molecular Weight (g/mol): 417.33
Molecular Formula: C25H18Cl2N2
InChi Key: CIZUMWWHWPJAAK-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(Cl)=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results