missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(4-allyloxyphenyl)fluorene 97.0+%, TCI America™

Catalog No. B46225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46225G 5 g
B462225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46225G Supplier TCI America Supplier No. B46225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142494-81-3
Molecular Formula C31H26O2
Molecular Weight (g/mol) 430.547
InChI Key OAGAWNXQZROGRJ-UHFFFAOYSA-N
PubChem CID 11604403
IUPAC Name 9,9-bis(4-prop-2-enoxyphenyl)fluorene
SMILES C=CCOC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OCC=C

Specifications

Melting Point 87°C
Color White
Quantity 5 g
Formula Weight 430.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-allyloxyphenyl)fluorene