accessibility menu, dialog, popup

UserName

H2COOCH2

CAS RN 142494-81-3

IUPAC Name: 9,9-bis[4-(prop-2-en-1-yloxy)phenyl]-9H-fluorene
Synonyms: 9,9-bis[4-(prop-2-en-1-yloxy)phenyl]fluorene
Molecular Weight (g/mol): 430.55
Molecular Formula: C31H26O2
InChi Key: OAGAWNXQZROGRJ-UHFFFAOYSA-N
SMILES: C=CCOC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC=C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results