missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Bifluorenylidene 98.0+%, TCI America™

Catalog No. B13181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13181G Supplier TCI America Supplier No. B13181G
Only null left

Chemical Identifiers

CAS 746-47-4
Molecular Formula C26H16
Molecular Weight (g/mol) 328.414
MDL Number MFCD00058940
InChI Key NVMYRXGXUBZAIU-UHFFFAOYSA-N
PubChem CID 136564
IUPAC Name 9-fluoren-9-ylidenefluorene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=C4C5=CC=CC=C5C6=CC=CC=C64

Specifications

Melting Point 187°C
Quantity 1 g
Formula Weight 328.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9′-Bifluorenylidene