CAS RN 746-47-4
IUPAC Name:
9,9'-bifluorenylidene
Synonyms:
9,9'-bifluorenylidene
Molecular Weight (g/mol):
328.41
Molecular Formula:
C26H16
InChi Key:
NVMYRXGXUBZAIU-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C1=CC=CC=C1C2=C1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
9,9'-Bifluorenylidene 98.0+%, TCI America™
CAS: 746-47-4 Molecular Formula: C26H16 Molecular Weight (g/mol): 328.414 MDL Number: MFCD00058940 InChI Key: NVMYRXGXUBZAIU-UHFFFAOYSA-N PubChem CID: 136564 IUPAC Name: 9-fluoren-9-ylidenefluorene SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3C2=C4C5=CC=CC=C5C6=CC=CC=C64