missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(4-Aminophenyl)carbazole hydrochloride, 98%
CAS: 312700-07-5 | C18H15ClN2 | 294.782 g/mol
Supplier: Thermo Scientific Chemicals L1136203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 9-(4-Aminophenyl)carbazole hydrochloride | |
| ∽250°C (decomposition) | |
| MFCD00082678 | |
| 4-9h-carbazol-9-yl aniline hydrochloride, 9-4-aminophenyl carbazole hydrochloride, 4-carbazol-9-yl aniline hydrochloride, 4-carbazol-9-ylaniline hydrochloride, acmc-1agoy | |
| C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N.Cl | |
| 294.782 | |
| 294.79 |
| 312700-07-5 | |
| C18H15ClN2 | |
| 1 g | |
| CFQROJKMLJRSAL-UHFFFAOYSA-N | |
| 4-carbazol-9-ylaniline;hydrochloride | |
| 2794996 | |
| 98% |
Chemical Identifiers
| 312700-07-5 | |
| 294.782 | |
| CFQROJKMLJRSAL-UHFFFAOYSA-N | |
| 2794996 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N.Cl |
| C18H15ClN2 | |
| MFCD00082678 | |
| 4-9h-carbazol-9-yl aniline hydrochloride, 9-4-aminophenyl carbazole hydrochloride, 4-carbazol-9-yl aniline hydrochloride, 4-carbazol-9-ylaniline hydrochloride, acmc-1agoy | |
| 4-carbazol-9-ylaniline;hydrochloride |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only